Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-2H-1,5-benzodioxepine-7-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. cc13203cb
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13203CB 250 mg Amber Glass Bottle
CC13203DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC13203CB Supplier Thermo Scientific Chemicals Supplier No. CC13203CB
Only null left
Add to Cart

Chemical Identifiers

CAS 321309-38-0
Molecular Formula C9H9ClO4S
Molecular Weight (g/mol) 248.68
MDL Number MFCD02681959
InChI Key ADAGPISZFDLGCR-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-sulfonyl chloride, 2h-1,5-benzodioxepin-7-sulfonylchloride, 3,4-dihydro, 2h-1,5-benzodioxepin-7-sulfonyl chloride, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepan-7-ylchlorosulfone, 3,4-dihydro-2h-1,5-benzodioxepin-7-sulfonyl chloride
PubChem CID 2776386
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC2=C(OCCCO2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 248.69
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepine-7-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.