Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxyallylbenzene, MP Biomedicals

Catalog No. ICN202038 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 g

Chemical Identifiers

CAS 93-15-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
InChI Key ZYEMGPIYFIJGTP-UHFFFAOYSA-N
Synonym methyleugenol, methyl eugenol, 4-allyl-1,2-dimethoxybenzene, eugenol methyl ether, 4-allylveratrole, o-methyleugenol, eugenyl methyl ether, 3,4-dimethoxyallylbenzene, methyl eugenol ether, veratrole methyl ether
PubChem CID 7127
ChEBI CHEBI:4918
IUPAC Name 1,2-dimethoxy-4-prop-2-enylbenzene
SMILES COC1=C(C=C(C=C1)CC=C)OC

Specifications

Quantity 100 g
Formula Weight 178.25
Chemical Name or Material 3, 4-Dimethoxyallylbenzene
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.