Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxybenzenesulfonyl chloride, 97%

Catalog No. AC354390050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 23095-31-0 | C8H9ClO4S | 236.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23095-31-0
Molecular Formula C8H9ClO4S
Molecular Weight (g/mol) 236.68
MDL Number MFCD00051769
InChI Key RSJSYCZYQNJQPY-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzene-1-sulfonyl chloride, 3,4-dimethoxybenzenesulfonylchloride, 3,4-dimethoxyphenylsulfonyl chloride, 3,4-dimethoxy benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethoxy, 3,4-dimethoxyphenyl chlorosulfone, pubchem5127, acmc-1co88, asischem d48920, zerenex zx009571
PubChem CID 2734183
IUPAC Name 3,4-dimethoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)S(=O)(=O)Cl)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 65.0°C to 70.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3SO2Cl
Quantity 5 g
Formula Weight 236.68
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dimethoxybenzenesulfonyl chloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.