Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethoxybenzylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
5763-61-1
C9H13NO2
167.21
MFCD00008116
DIVNUTGTTIRPQA-UHFFFAOYSA-N
3,4-dimethoxybenzylamine,veratrylamine,3,4-dimethoxyphenyl methanamine,benzenemethanamine, 3,4-dimethoxy,1-3,4-dimethoxyphenyl methanamine,benzylamine, 3,4-dimethoxy,3,4-dimethoxyphenyl methylamine,veratrylamin,pubchem7394,acmc-1altc
79832
(3,4-dimethoxyphenyl)methanamine
COC1=C(C=C(C=C1)CN)OC
5763-61-1
C9H13NO2
167.21
MFCD00008116
DIVNUTGTTIRPQA-UHFFFAOYSA-N
3,4-dimethoxybenzylamine,veratrylamine,3,4-dimethoxyphenyl methanamine,benzenemethanamine, 3,4-dimethoxy,1-3,4-dimethoxyphenyl methanamine,benzylamine, 3,4-dimethoxy,3,4-dimethoxyphenyl methylamine,veratrylamin,pubchem7394,acmc-1altc
79832
(3,4-dimethoxyphenyl)methanamine
COC1=C(C=C(C=C1)CN)OC
Specific Gravity 1.1
Flash Point >112°C
Boiling Point 281°C to 284°C
Refractive Index 1.5530 to 1.557
Density 1.1000g/mL
Color Yellow
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (CH3O)2C6H3CH2NH2
Beilstein 13, 796
Formula Weight 167.21
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3, 4-Dimethoxybenzylamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 227-287-7