Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Di-n-butyl ether, 99+%, Extra Dry, AcroSeal™
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -95.0°C |
Color | Colorless |
Density | 0.7680g/mL |
Boiling Point | 141.0°C |
Flash Point | 25°C |
Assay Percent Range | 99% min. (GC) |
Packaging | AcroSeal glass Bottle |
Refractive Index | 1.3978 to 1.3998 |
Linear Formula | [CH3(CH2)3]2O |
Quantity | 1 L |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful to aquatic life with long lasting effects. May cause respiratory irritation. Causes skin irritation. Flammable liquid and vapor. |
Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN |
EINECSNumber | 205-575-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction