Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichloro-1,2,4-thiadiazole, 97%

Catalog No. AC376030050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 2254-88-8 | C2Cl2N2S | 155.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2254-88-8
Molecular Formula C2Cl2N2S
Molecular Weight (g/mol) 155.01
InChI Key CEUHPOVLEQUFCC-UHFFFAOYSA-N
PubChem CID 2725055
IUPAC Name 3,5-dichloro-1,2,4-thiadiazole
SMILES C1(=NSC(=N1)Cl)Cl

Specifications

Color Colorless to Yellow
Boiling Point 165.0°C to 167.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5580 to 1.5600
Quantity 5 g
Synonym dichloro-1,2,4-thiadiazole, 3,5-dichloro-1,2,4 thiadiazole, 1,2,4-thiadiazole, 3,5-dichloro, 1,2,4-thiadiazole,3,5-dichloro, 3,5-dichloro-1,2,4 thiadiazol
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 155.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3,5-Dichloro-1,2,4-thiadiazole
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.