Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorobenzyl alcohol, 98%, Thermo Scientific™

Catalog No. p-104568
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC182770050 5 g Glass bottle
AC182770250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC182770050 Supplier Thermo Scientific Chemicals Supplier No. 182770050
Only null left
Add to Cart

Chemical Identifiers

CAS 60211-57-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.03
MDL Number MFCD00004634
InChI Key VSNNLLQKDRCKCB-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzyl alcohol, 3,5-dichlorophenyl methanol, benzenemethanol, 3,5-dichloro, 3,5-dichlorobenzylalcohol, 3,5-dichlorobenzylic alcohol, 3,5-dichlorobenzenemethanol, 3,5-dicholorbenzylalcohol, 3,5-dichlorophenyl methan-1-ol, 3, 5-dichlorobenzyl alcohol, pubchem4908
PubChem CID 43236
IUPAC Name (3,5-dichlorophenyl)methanol
SMILES C1=C(C=C(C=C1Cl)Cl)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 76°C to 80°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3CH2OH
Quantity 5 g
Formula Weight 177.03
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dichlorobenzyl alcohol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 262-109-1
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.