Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorobenzylamine, 95%, Thermo Scientific™

Catalog No. TL00168EA
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 39989-43-0
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00052681
InChI Key ICIJWOWQUHHETJ-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzylamine, 3,5-dichlorophenyl methanamine, benzenemethanamine, 3,5-dichloro, 3,5-dichloro-benzylamine, 1-3,5-dichlorophenyl methanamine, 3,5-dichlorophenyl methylamine, pubchem23431, 3.5-dichlorobenzylamine, acmc-1cthv, integrase inhibitor, r1 4
PubChem CID 457602
SMILES NCC1=CC(Cl)=CC(Cl)=C1

Specifications

CAS Min % 95.0
Density 1.3200g/mL
Assay Percent Range 95%
Linear Formula Cl2C6H3CH2NH2
Quantity 10 g
IUPAC Name 1-(3,5-dichlorophenyl)methanamine
Formula Weight 176.05
Percent Purity 95%
Chemical Name or Material 3, 5-Dichlorobenzylamine
Hazard Category Danger
Hazard Statement GHS P Statement Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.