Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dihydroxybenzaldehyde, 98%

Catalog No. AC425650050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425650050 5 g Glass bottle
AC425650010 1 g Glass bottle
AC425650250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC425650050 Supplier Thermo Scientific Chemicals Supplier No. 425650050
Only null left
Add to Cart

CAS: 26153-38-8 | C7H6O3 | 138.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26153-38-8
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00016611
InChI Key HAQLHRYUDBKTJG-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehyde
PubChem CID 94365
ChEBI CHEBI:50204
IUPAC Name 3,5-dihydroxybenzaldehyde
SMILES OC1=CC(C=O)=CC(O)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 153.0°C to 158.0°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)2C6H3CHO
Quantity 5 g
Formula Weight 138.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dihydroxybenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye prote
EINECSNumber 247-479-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.