Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde, 99%, Thermo Scientific™

Catalog No. AC408520050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408520050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC408520050 Supplier Thermo Scientific Chemicals Supplier No. 408520050
Only null left
Add to Cart

Chemical Identifiers

CAS 22042-79-1
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD00051396
InChI Key VOMRTQQGXWPTJK-UHFFFAOYSA-N
PubChem CID 89179
IUPAC Name 3,5-dimethyl-1-phenylpyrazole-4-carbaldehyde
SMILES CC1=C(C(=NN1C2=CC=CC=C2)C)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 125°C to 128°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbaldehyde, 3,5-dimethyl-1-phenylpyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 3,5-dimethyl-1-phenyl, 3,5-dimethyl-1-phenyl-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3,5-dimethyl-1-phenyl, 3,5-dimethyl-4-formyl-1-phenylpyrazole, 3,5-dimethyl-1-phenyl-4-pyrazolecarboxaldehyde, 3?5-dimethyl-1-phenylpyrazole-4-carboxaldehyde, 3,5-dimethyl-1-phenyl 1h-pyrazole-4-carbaldehyde, 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbaldehyde #
Formula Weight 200.24
Percent Purity 99%
Chemical Name or Material 3, 5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde, 99%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 244-750-9
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.