Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dichloropyridazine-4-carboxylic acid, 98%, Thermo Scientific™

Catalog No. AAL1041803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1041803 1 g
AAL10418MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1041803 Supplier Thermo Scientific Chemicals Supplier No. L1041803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51149-08-7
Molecular Formula C5H2Cl2N2O2
Molecular Weight (g/mol) 192.98
MDL Number MFCD00011575
InChI Key FRCXPDWDMAYSCE-UHFFFAOYSA-N
Synonym 3,6-dichloro-4-pyridazinecarboxylic acid, 3,6-dichloropyridazine-4-carboxylicacid, 4-pyridazinecarboxylic acid, 3,6-dichloro, 3,6-dichloropyridazinecarboxylic acid, 3,6-dichloro-pyridazine-4-carboxylic acid, pubchem9529, enamine_001708, acmc-1ay8l, ksc269c3r, 4-carboxy-3,6-dichloropyridazine
PubChem CID 433804
IUPAC Name 3,6-dichloropyridazine-4-carboxylic acid
SMILES OC(=O)C1=CC(Cl)=NN=C1Cl

Specifications

Melting Point 137°C to 140°C
Quantity 1 g
Beilstein 610056
Formula Weight 192.99
Percent Purity 98%
Chemical Name or Material 3,6-Dichloropyridazine-4-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.