Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,8-Diamino-6-phenylphenanthridine, 98%

Catalog No. AAB2404906
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 52009-64-0 | C19H15N3 | 285.35 g/mol

3,8-Diamino-6-phenylphenanthridine is used in the synthesis of rigid polyamides. It has anti-tumor and anti-viral properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,8-Diamino-6-phenylphenanthridine is used in the synthesis of rigid polyamides. It has anti-tumor and anti-viral properties.

Solubility
Soluble in chloroform, dichloromethane and methanol.

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52009-64-0
Molecular Formula C19H15N3
Molecular Weight (g/mol) 285.35
MDL Number MFCD00075152
InChI Key CPNAVTYCORRLMH-UHFFFAOYSA-N
Synonym 3,8-diamino-6-phenylphenanthridine, 6-phenylphenathridine-3,8-diamine, 3,8-dapp, 3,8-diamino-6-phenyl-phenanthridine, acmc-1aycj, 6-phenyl-3,8-phenanthridinediamine, 3,8-diamino-6-phenyl phenanthridine, 3,8-phenanthridinediamine,6-phenyl, 3,8-diamino-6-phenylphenanthridine6-phenyl-3,8-phenanthridinediamine,
PubChem CID 104052
IUPAC Name 6-phenylphenanthridine-3,8-diamine
SMILES C1=CC=C(C=C1)C2=C3C=C(C=CC3=C4C=CC(=CC4=N2)N)N

Specifications

Melting Point 196°C to 198°C
Quantity 5 g
Solubility Information Soluble in chloroform,dichloromethane and methanol.
Formula Weight 285.35
Percent Purity 98%
Chemical Name or Material 3,8-Diamino-6-phenylphenanthridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 257-602-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.