Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-hydroxyphenyl Methyl Sulfone 97.0+%, TCI America™
SDP

Catalog No. A219825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A219825G 25 g
A21985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A219825G Supplier TCI America Supplier No. A219825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 98-30-6
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.21
MDL Number MFCD00719450
InChI Key SFLMBHYNCSYPOO-UHFFFAOYSA-N
Synonym 2-Amino-4-(methylsulfonyl)phenol, 2-Hydroxy-5-(methylsulfonyl)aniline
PubChem CID 7382
IUPAC Name 2-amino-4-methanesulfonylphenol
SMILES CS(=O)(=O)C1=CC=C(O)C(N)=C1

Specifications

Melting Point 158°C
Quantity 25 g
Formula Weight 187.21
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-hydroxyphenyl Methyl Sulfone
EINECSNumber (3)-2140
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.