Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-6-(trifluoromethyl)-1H-indazole, 95%, Thermo Scientific™

Catalog No. p-4228348
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435810010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435810010 Supplier Thermo Scientific Chemicals Supplier No. 435810010

Chemical Identifiers

CAS 2250-55-7
Molecular Formula C8H6F3N3
Molecular Weight (g/mol) 201.15
MDL Number MFCD03426695
InChI Key ZMPFPQFXUYYHSJ-UHFFFAOYSA-N
Synonym 6-trifluoromethyl-1h-indazol-3-amine, 3-amino-6-trifluoromethyl-1h-indazole, trifluoromethylindazolamine, 3-amino-6-trifluoromethylindazole, 6-trifluoro methyl-1h-indazole-3-amine, 6-trifluoromethyl-1h-indazole-3-amine, 1h-indazol-3-amine,6-trifluoromethyl, 6-trifluoromethyl-1h-indazol-3-ylamine, 6-trifluoromethyl-1h-indazole-3-ylamine
PubChem CID 817909
IUPAC Name 6-(trifluoromethyl)-1H-indazol-3-amine
SMILES C1=CC2=C(C=C1C(F)(F)F)NN=C2N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 173.0°C to 175.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 201.15
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-6-(trifluoromethyl)-1H-indazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.