Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminoazetidine dihydrochloride, 95%

Catalog No. aah59318md
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59318MD 250 mg
AAH59318ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59318MD Supplier Thermo Scientific Chemicals Supplier No. H59318MD
Only null left
Add to Cart

CAS: 102065-89-4 | C3H8N2 | 72.111 g/mol

3-Aminoazetidine dihydrochloride, is used as active pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminoazetidine dihydrochloride, is used as active pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102065-89-4
Molecular Formula C3H8N2
Molecular Weight (g/mol) 72.111
MDL Number MFCD09910173
InChI Key FDPKMJDUXJFKOI-UHFFFAOYSA-N
Synonym 3-aminoazetidine, 3-azetidinamine, acmc-209y1n, 3-aminoazetidine dihy drochloric
PubChem CID 1516506
IUPAC Name azetidin-3-amine
SMILES C1C(CN1)N

Specifications

Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 145.03
Percent Purity 95%
Chemical Name or Material 3-Aminoazetidine dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.