Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzenesulfonamide, 97+%

Catalog No. AAB2185714
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2185714

Supplier: Thermo Scientific Chemicals B2185714

Only null left
Add to Cart

CAS: 98-18-0 | C6H8N2O2S | 172.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-18-0
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00035781
InChI Key JPVKCHIPRSQDKL-UHFFFAOYSA-N
Synonym metanilamide, m-aminobenzenesulfonamide, benzenesulfonamide, 3-amino, m-sulfamoylaniline, 3-aminobenzene-1-sulfonamide, benzenesulfonamide, m-amino, m-aminobenzenesulphonamide, metaniilamide, 3-aminosulfonylaniline, 3-amino-benzenesulfonamide
PubChem CID 7377
IUPAC Name 3-aminobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)N

Specifications

Melting Point 139°C to 142°C
Quantity 25 g
Formula Weight 172.21
Percent Purity ≥97%
Chemical Name or Material 3-Aminobenzenesulfonamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-646-0
RTECSNumber OY2200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.