Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzenesulfonyl Fluoride Hydrochloride 98%, Thermo Scientific™

Catalog No. AC159930050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC159930050 5 g
AC159930250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159930050 Supplier Thermo Scientific Chemicals Supplier No. 159930050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3887-48-7
Molecular Formula C6H7ClFNO2S
Molecular Weight (g/mol) 211.64
MDL Number MFCD00012972
InChI Key KTADZFWRDLKHKW-UHFFFAOYSA-N
Synonym 3-aminobenzenesulfonyl fluoride hydrochloride, metanilyl fluoride, hydrochloride, benzenesulfonyl fluoride, 3-amino-, hydrochloride, benzenesulfonyl fluoride, 3-amino-, hydrochloride 1:1, metanilyl fluoride hcl, metanilyl fluoride hydrochloride, m-aminobenzenesulphonyl fluoride hydrochloride, 3-aminophenyl fluorosulfone, chloride, 3-aminobenzenesulfonylfluoride hydrochloride, 3-aminophenylsulfonyl fluoride,hydrochloride
PubChem CID 77496
IUPAC Name hydrogen 3-aminobenzene-1-sulfonyl fluoride chloride
SMILES [H+].[Cl-].NC1=CC=CC(=C1)S(F)(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown to Pink
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula H2NC6H4SO2F·HCl
Quantity 5 g
Beilstein 14, II, 434
Formula Weight 211.64
Percent Purity ≥97.5%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Aminobenzenesulfonyl fluoride hydrochloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-429-7
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.