Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzylamine, 97%

Catalog No. p-3586352
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 4403-70-7 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4403-70-7
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00078355
InChI Key ZDBWYUOUYNQZBM-UHFFFAOYSA-N
Synonym 3-aminobenzylamine, 3-aminomethyl aniline, m-aminobenzylamine, benzenemethanamine, 3-amino, 3-aminomethyl-aniline, pubchem5802, alpha-amino-m-toluidine, 3-aminomethyl phenylamine, 3-aminophenyl methylamine
PubChem CID 4628831
IUPAC Name 3-(aminomethyl)aniline
SMILES NCC1=CC(N)=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 36°C to 39°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula NH2CH2C6H4NH2
Quantity 5 g
Formula Weight 122.17
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 3-Aminobenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 412-230-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.