Learn More
3-(Aminomethyl)benzoic acid hydrochloride, 95%, Thermo Scientific Chemicals
Description
It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 242°C to 244°C |
Quantity | 250 mg |
Beilstein | 3697656 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 187.62 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 3-(Aminomethyl)benzoic acid hydrochloride |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
TSCA | No |
storageNote1 | 2° to 8°C |
Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.