Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Aminopropyl)triethoxysilane, 98%

Catalog No. AAA1066836
Click to view available options
Quantity:
100 g
500 g
2500 g

Alfa Aesar (3-Aminopropyl)triethoxysilane, 98%, Quantity: 100g, Boiling Point: 217 deg.C, CAS: 919-30-2, Molecular Formula: C9H23NO3Si, Molecular Weight: 221.372, Percent Purity: 98%, pH: 11 | CAS: 919-30-2 | C9H23NO3Si | 221.37 g/mol

(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)triethoxysilane is useful in the formation of aminopropyl derivative of glass. It acts as an adsorbent for affinity chromatography. It is used to prepare positively charged slides suitable for use with various immunohistochemical and in situ hybridization procedures. In addition, it is used as adhesives and sealant chemicals, paint additives and coating additives. Further, it is used as a silylation reagent for coating glass and silica surfaces and to crosslink and immobilize proteins and other molecules.

Solubility
Miscible with toluene, acetone, chloroform and ethanol.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, acids and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 919-30-2
Molecular Formula C9H23NO3Si
Molecular Weight (g/mol) 221.37
MDL Number MFCD00008207,MFCD01324904
InChI Key WYTZZXDRDKSJID-UHFFFAOYSA-N
Synonym 3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
PubChem CID 13521
IUPAC Name 3-triethoxysilylpropan-1-amine
SMILES CCO[Si](CCCN)(OCC)OCC

Specifications

Boiling Point 217°C
pH 11
Quantity 500 g
Linear Formula H2N(CH2)3Si(OCH2CH3)3
UN Number UN2735
Beilstein 1754988
Flash Point 104°C (219°F)
Solubility Information Miscible with toluene,acetone,chloroform and ethanol.
Formula Weight 221.37
Percent Purity 98%
Odor Amine-like
Refractive Index 1.421
Sensitivity Air and moisture sensitive
Density 0.948
Chemical Name or Material (3-Aminopropyl)triethoxysilane
Show More Show Less
Hazard Category H302-H314-H317-H335
Hazard Statement GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 213-048-4
RTECSNumber TX2100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.