Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminotetrahydrofuran 97.0+%, TCI America™
SDP

Catalog No. p-7118030
Click to view available options
Quantity:
250 mg
1 g

Chemical Identifiers

CAS 88675-24-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD07778394
InChI Key MIPHRQMEIYLZFZ-UHFFFAOYSA-N
PubChem CID 3365553
IUPAC Name oxolan-3-amine
SMILES C1COCC1N

Specifications

Color Colorless
Boiling Point 73°C
Quantity 250 mg
UN Number 1993
Formula Weight 87.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Aminotetrahydrofuran
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.