Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzylamino-1-propanol 98.0+%, TCI America™
SDP

Catalog No. B346225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B346225G 25 g
B34625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B346225G Supplier TCI America Supplier No. B346225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4720-29-0
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.236
MDL Number MFCD00020622
InChI Key ZQJXSIOFSZYGMH-UHFFFAOYSA-N
Synonym 3-benzylamino propan-1-ol, n-benzyl-3-aminopropan-1-ol, 3-benzylamino-1-propanol, 3-benzylamino propanol, 1-propanol,3-methylphenylamino, 1-propanol, 3-phenylmethyl amino, zlchem 680, 3-benzylaminopropanol
PubChem CID 78448
IUPAC Name 3-(benzylamino)propan-1-ol
SMILES C1=CC=C(C=C1)CNCCCO

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
Formula Weight 165.24
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3-Benzylamino-1-propanol
EINECSNumber (3)-0314
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.