Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Boc-amino)phenol, 97%

Catalog No. p-7036719
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 19962-06-2 | C11H15NO3 | 209.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19962-06-2
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD06798070
InChI Key HJQNVUQTARSZDK-UHFFFAOYSA-N
Synonym tert-butyl 3-hydroxyphenylcarbamate, 3-boc-amino phenol, tert-butyl n-3-hydroxyphenyl carbamate, tert-butyl 3-hydroxyphenyl carbamate, n-boc-3-aminophenol, carbamic acid, 3-hydroxyphenyl-, 1,1-dimethylethyl ester, carbamic acid, n-3-hydroxyphenyl-, 1,1-dimethylethyl ester, carbamic acid,n-3-hydroxyphenyl-, 1,1-dimethylethyl ester, tert-butoxy-n-3-hydroxyphenyl carboxamide, n-boc-3-amino-phenol
PubChem CID 312485
IUPAC Name tert-butyl N-(3-hydroxyphenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=CC(O)=C1

Specifications

Melting Point 134°C to 138°C
Quantity 1 g
Formula Weight 209.25
Percent Purity 97%
Chemical Name or Material 3-(Boc-amino)phenol
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.