Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 96%, Thermo Scientific™

Catalog No. AC301940010
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 76824-99-2
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD03092912
InChI Key CRAGDYRHPWTZJL-UHFFFAOYNA-N
Synonym 6-hydroxy-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, akos 68, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, 6-hydroxy-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid h2o, 6-oxidanyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate, d,l-6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
PubChem CID 18525693
SMILES OC(=O)C1CC2=C(CN1)C=CC(O)=C2

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 270°C to 276°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 1 g
IUPAC Name 6-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Formula Weight 211.22
Percent Purity ≥95%
Physical Form Crystalline Powder
Chemical Name or Material DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.