Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-o-xylene, 99%, Thermo Scientific™

Catalog No. ac107580050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 576-23-8
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000069
InChI Key WLPXNBYWDDYJTN-UHFFFAOYSA-N
Synonym 3-bromo-o-xylene, 3-bromo-1,2-dimethylbenzene, 2,3-dimethylbromobenzene, 2,3-dimethyl bromobenzene, benzene, bromodimethyl, 1-bromo-2,3-dimethyl-benzene, benzene, 1-bromo-2,3-dimethyl, bromoxilene, xylylbromid, bromo-o-xylene
PubChem CID 68472
IUPAC Name 1-bromo-2,3-dimethylbenzene
SMILES CC1=CC=CC(Br)=C1C

Specifications

Color Colorless
CAS Min % 98.5
CAS Max % 100.0
Density 1.3600g/mL
Boiling Point 214°C
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5590 to 1.561
Assay Percent Range 99%
Specific Gravity 1.36
Linear Formula (CH3)2C6H3Br
Beilstein 05, 365
Formula Weight 185.06
Percent Purity 99%
Chemical Name or Material 3-Bromo-o-xylene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-398-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.