Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenyl Methyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. B389325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B389325G 25 g
B38935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B389325G Supplier TCI America Supplier No. B389325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34896-80-5
Molecular Formula C7H7BrO2S
Molecular Weight (g/mol) 235.10
MDL Number MFCD03095326
InChI Key WBOMXUMQOVQNKT-UHFFFAOYSA-N
PubChem CID 2757146
IUPAC Name 1-bromo-3-methanesulfonylbenzene
SMILES CS(=O)(=O)C1=CC=CC(Br)=C1

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 25 g
Formula Weight 235.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromophenyl Methyl Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.