Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromopyrrolidine hydrobromide, 96%, Thermo Scientific Chemicals

Catalog No. AAH66833MD
Click to view available options
Quantity:
250 mg
1 g

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Notes
Moisture and Light Sensitive, store in dark and away form water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1262769-75-4
Molecular Formula C4H9Br2N
Molecular Weight (g/mol) 230.931
MDL Number MFCD13181592
InChI Key ZSDJSWROXIUGAE-UHFFFAOYSA-N
Synonym 3-bromopyrrolidine hydrobromide, 3-bromopyrrolidine hbr, 3-bromopyrrolidine-hydrogen bromide 1/1
PubChem CID 66570691
IUPAC Name 3-bromopyrrolidine;hydrobromide
SMILES C1CNCC1Br.Br

Specifications

Melting Point 135°C to 141°C
Color Brown to Pink
Quantity 250 mg
UN Number UN3335
Sensitivity Light and moisture sensitive
Formula Weight 230.93
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Bromopyrrolidine hydrobromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.