Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-methoxypyridine-4-boronic acid, 95%, Thermo Scientific™

Catalog No. AC448770010
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448770010 1 g Glass bottle
AC448772500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448770010 Supplier Thermo Scientific Chemicals Supplier No. 448770010
Add to Cart

Chemical Identifiers

CAS 957060-88-7
Molecular Formula C6H7BClNO3
Molecular Weight (g/mol) 187.39
MDL Number MFCD06798255
InChI Key YQNAXOGPSUVHNU-UHFFFAOYSA-N
Synonym 3-chloro-2-methoxypyridine-4-boronic acid, 3-chloro-2-methoxypyridin-4-yl boronic acid, 3-chloro-2-methoxypyridin-4-ylboronicacid, 2-methoxy-3-chloropyridine-4-boronic acid, 3-chloro-2-methoxy-4-pyridineboronic acid, pubchem17058, abbypharma ap-17-5909, 3-chloro-2-methoxypyrdine-4-boronic acid, boronic acid, b-3-chloro-2-methoxy-4-pyridinyl
PubChem CID 44119809
IUPAC Name (3-chloro-2-methoxypyridin-4-yl)boronic acid
SMILES COC1=NC=CC(B(O)O)=C1Cl

Specifications

Melting Point 140°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 187.39
Percent Purity 95%
Chemical Name or Material 3-Chloro-2-methoxypyridine-4-boronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.