Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-(pentafluorothio)benzonitrile, 97%, Thermo Scientific™

Catalog No. AAH33840MD
Click to view available options
Quantity:
250 mg

Catalog No. AAH33840MD

Supplier: Thermo Scientific Chemicals H33840MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159727-28-3
Molecular Formula C7H3ClF5NS
Molecular Weight (g/mol) 263.61
MDL Number MFCD22201047
InChI Key HASUULAKVNPSHK-UHFFFAOYSA-N
Synonym 3-chloro-5-pentafluorosulfur benzonitrile, 3-chloro-5-pentafluoro-??-sulfanyl benzonitrile, 3-chloro-5-pentafluorothio benzonitrile
PubChem CID 86277648
SMILES FS(F)(F)(F)(F)C1=CC(Cl)=CC(=C1)C#N

Specifications

Quantity 250 mg
UN Number UN3439
IUPAC Name 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzonitrile
Formula Weight 263.62
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(pentafluorothio)benzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.