Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-6-methoxypyridazine 97.0+%, TCI America™
SDP

Catalog No. C237625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C237625G 25 g
C23765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C237625G Supplier TCI America Supplier No. C237625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1722-10-7
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.56
MDL Number MFCD00006467
InChI Key XBJLKXOOHLLTPG-UHFFFAOYSA-N
PubChem CID 74403
IUPAC Name 3-chloro-6-methoxypyridazine
SMILES COC1=CC=C(Cl)N=N1

Specifications

Melting Point 91°C
Color White-Yellow
Boiling Point 175°C
Quantity 25 g
Synonym pyridazine, 3-chloro-6-methoxy, 3-chloro-6-methoxy-pyridazine, 6-chloro-3-methoxypyridazine, pyridazine,3-chloro-6-methoxy, 6-chloropyridazin-3-yl methyl ether, 3-chloro-6-methoxypyridazine, zlchem 731, acmc-1bvtl, 3-methoxy-6-chloropyridazine, 3-chloro-6-methoxy pyridazine
Formula Weight 144.56
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-6-methoxypyridazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.