Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropyl isocyanate, 96%, Thermo Scientific™

Catalog No. AC314590050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314590050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC314590050 Supplier Thermo Scientific Chemicals Supplier No. 314590050

Chemical Identifiers

CAS 13010-19-0
Molecular Formula C4H6ClNO
Molecular Weight (g/mol) 119.55
MDL Number MFCD00010384
InChI Key RQAVSDINDRNIKL-UHFFFAOYSA-N
Synonym 3-chloropropyl isocyanate, 3-chloropropylisocyanate, propane, 1-chloro-3-isocyanato, 1-chloro-3-isocyanato-propane, chloropropylisocyanate, chloropropyl isocyanate, 3-chloropropanisocyanate, chloro propyl isocyanate, acmc-20an5q, 3-chloropropyl-isocyanate
PubChem CID 166752
IUPAC Name 1-chloro-3-isocyanatopropane
SMILES ClCCCN=C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Density 1.1600g/mL
Boiling Point 152°C to 154°C
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.453
Linear Formula Cl(CH2)3NCO
Quantity 5 g
Specific Gravity 1.16
Formula Weight 119.55
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Chloropropyl isocyanate, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.