Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Chlorosulfonyl)benzoyl chloride, 98%

Catalog No. AC405070250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

Catalog No. AC405070250

Supplier: Thermo Scientific Chemicals 405070250

Only null left
Add to Cart

CAS: 4052-92-0 | C7H4Cl2O3S | 239.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4052-92-0
Molecular Formula C7H4Cl2O3S
Molecular Weight (g/mol) 239.07
MDL Number MFCD00051352
InChI Key XWEBTVZIZWEJOO-UHFFFAOYSA-N
Synonym 3-chlorosulfonyl benzoyl chloride, benzoyl chloride, 3-chlorosulfonyl, m-chlorosulfonyl benzoyl chloride, 3-chlorosulphonyl benzoyl chloride, 3-chlorosulphonylbenzoyl chloride, 3-chlorosulfonyl-benzoyl chloride, benzoyl chloride,3-chlorosulfonyl, benzoyl chloride, m-chlorosulfonyl-7ci,8ci, c7h4cl2so3, 3-chlorosulfonylbenzoyl=chloride
PubChem CID 77671
IUPAC Name 3-chlorosulfonylbenzoyl chloride
SMILES ClC(=O)C1=CC=CC(=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 18°C to 20°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5780 to 1.58
Linear Formula COClC6H4SO2Cl
Quantity 25 g
Formula Weight 239.08
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 3-(Chlorosulfonyl)benzoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-759-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.