Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclopropylpropargyl diethyl acetal, 97%, Thermo Scientific™

Catalog No. AC391650050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 146195-69-9
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD12912455
InChI Key WOSJTIAXBXVXLF-UHFFFAOYSA-N
Synonym 3-cyclopropylpropiolaldehyde diethyl acetal, 3,3-diethoxyprop-1-yn-1-yl cyclopropane, 3-cyclopropylpropargyl diethyl acetal, 3-cyclopropylpropargyl aldehyde diethyl acetal
PubChem CID 42580279
IUPAC Name 3,3-diethoxyprop-1-ynylcyclopropane
SMILES CCOC(C#CC1CC1)OCC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9700g/mL
Boiling Point 236°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4520 to 1.4540 (20°C, 589nm)
Linear Formula C3H5CCCH(OCH2CH3)2
Quantity 5 g
Specific Gravity 0.97
Formula Weight 168.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Cyclopropylpropargyl diethyl acetal
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.