Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Di-tert-butylphosphonium)propane sulfonate, 97%

Catalog No. p-7032693
Click to view available options
Quantity:
250 mg
1 g

CAS: 1055888-89-5 | C11H25O3PS | 268.352 g/mol

3-(Di-tert-butylphosphino)propane-1-sulfonic acid is used in palladium catalyzed cross-coupling reactions. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Di-tert-butylphosphino)propane-1-sulfonic acid is used in palladium catalyzed cross-coupling reactions. It is also used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Moisture and air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1055888-89-5
Molecular Formula C11H25O3PS
Molecular Weight (g/mol) 268.352
MDL Number MFCD12546062
InChI Key JPNPRWMRUCIEMN-UHFFFAOYSA-N
Synonym 3-di-t-butylphosphonium propane sulfonate, 3-di-tert-butylphosphino propane-1-sulfonic acid, dtbpps, di-t-butyl 3-sulfonatopropyl phosphine, 3-di-tert-butylphosphonium propane sulfonate, 3-di-tert-butylphosphanyl propane-1-sulfonic acid
PubChem CID 24949188
IUPAC Name 3-ditert-butylphosphanylpropane-1-sulfonic acid
SMILES CC(C)(C)P(CCCS(=O)(=O)O)C(C)(C)C

Specifications

Linear Formula [(CH3)3C]2P(CH2)3SO3H
Quantity 250 mg
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 268.35
Percent Purity 97%
Chemical Name or Material 3-(Di-tert-butylphosphonium)propane sulfonate
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.