Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxypropionic Acid 98.0+%, TCI America™
SDP

Catalog No. E005325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E005325ML 25 mL
E00535ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E005325ML Supplier TCI America Supplier No. E005325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4324-38-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00058997
InChI Key JRXXEXVXTFEBIY-UHFFFAOYSA-N
Synonym O-Ethylhydracrylic Acid
PubChem CID 61351
IUPAC Name 3-ethoxypropanoic acid
SMILES CCOCCC(=O)O

Specifications

Color Colorless
Boiling Point 219°C
Quantity 25 mL
Formula Weight 118.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Ethoxypropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.