Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Fluoro-2-formylbenzeneboronic acid, 95%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Undergoes unprecedented tautomeric rearrangement in solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Undergoes unprecedented tautomeric rearrangement in solution.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Undergoes unprecedented tautomeric rearrangement in solution.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 123°C to 127°C |
| Quantity | 25 g |
| UN Number | UN3261 |
| Sensitivity | Light sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 167.93 |
| Percent Purity | 95% |
| Physical Form | Solid |
| Chemical Name or Material | 3-Fluoro-2-formylbenzeneboronic acid |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction