Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hexyn-1-ol, 98%, Thermo Scientific™

Catalog No. AC223480050
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC223480050 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC223480050 Supplier Thermo Scientific Chemicals Supplier No. 223480050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1002-28-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00002957
InChI Key ODEHKVYXWLXRRR-UHFFFAOYSA-N
Synonym 3-hexyn-1-ol, 3-hexynol, 3-hexyne-1-ol, unii-x07kyi4gth, x07kyi4gth, acmc-2097nm, 3-hexyn-1-ol, 98 %, 3-hexyn-1-ol 5ml
PubChem CID 66083
IUPAC Name hex-3-yn-1-ol
SMILES CCC#CCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.8900g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4540 to 1.4560 (20°C, 589nm)
Linear Formula C2H5Cidentical toCCH2CH2OH
Quantity 5 mL
Beilstein 01, III, 1986
Formula Weight 98.14
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 3-Hexyn-1-ol, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.