Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-1-methyl-5,6-indolinedione, 98%

Catalog No. p-4089374
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426210010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426210010 Supplier Thermo Scientific Chemicals Supplier No. 426210010

CAS: 54-06-8 | C9H9NO3 | 179.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-06-8
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.17
MDL Number MFCD00069732
InChI Key RPHLQSHHTJORHI-UHFFFAOYSA-N
Synonym adrenochrome, adraxone, usaf uctl-7, 3-hydroxy-1-methyl-5,6-indolinedione, 1-adrenochrome, 3-hydroxy-1-methylindoline-5,6-dione, 2,3-dihydro-3-hydroxy-1-methyl-1h-indole-5,6-dione, 5,6-indolinedione, 3-hydroxy-1-methyl, adrenochrome, l, adrenochrom
PubChem CID 5898
IUPAC Name 3-hydroxy-1-methyl-2,3-dihydroindole-5,6-dione
SMILES CN1CC(C2=CC(=O)C(=O)C=C21)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 161
Formula Weight 179.17
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-1-methyl-5, 6-indolinedione
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-192-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.