Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-phenyl-2-butanone, 96%

Catalog No. aal0717606
Click to view available options
Quantity:
5 g

CAS: 2893-05-2 | C6H15NO | 117.19 g/mol

3-Methyl-1-phenyl-2-butanone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-butanone is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2893-05-2
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00040487
InChI Key VHYUNSUGCNKWSO-UHFFFAOYSA-N
Synonym 3-isopropoxypropylamine, 3-isopropoxy-1-propylamine, isopropoxypropylamine, 1-propanamine, 3-1-methylethoxy, 3-aminopropyl isopropyl ether, 3-1-methylethoxy-1-propanamine, 3-isopropoxy-n-propylamine, 3-isopropoxypropan-1-amine, propylamine, 3-isopropoxy, 3-propan-2-yloxy propan-1-amine
PubChem CID 76194
IUPAC Name 3-propan-2-yloxypropan-1-amine
SMILES CC(C)OCCCN

Specifications

Density 0.968
Boiling Point 237°C to 240°C
Refractive Index 1.504
Quantity 5 g
Beilstein 2042880
Solubility Information Not miscible in water.
Formula Weight 162.23
Percent Purity 96%
Chemical Name or Material 3-Methyl-1-phenyl-2-butanone
EINECSNumber 220-765-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.