Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-1-propanol, 98%

Catalog No. p-4259005
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

CAS: 1589-49-7 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1589-49-7
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key JDFDHBSESGTDAL-UHFFFAOYSA-N
Synonym 3-methoxy-1-propanol, 1-propanol, 3-methoxy, 3-methoxypropanol, trimethylene glycol monomethyl ether, beta-pgme, 1,3-propanediol monomethyl ether, unii-oak281wz1p, propylene glycol monomethyl ether, beta, oak281wz1p, mpol
PubChem CID 74116
IUPAC Name 3-methoxypropan-1-ol
SMILES COCCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9420g/mL
Boiling Point 149.0°C to 150.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4120 to 1.4140 (20°C, 589nm)
Linear Formula CH3O(CH2)3OH
Quantity 10 g
Specific Gravity 0.942
Formula Weight 90.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxy-1-propanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.