Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-2-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Catalog No. AAH3397203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3397203 1 g
AAH33972MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3397203 Supplier Thermo Scientific Chemicals Supplier No. H3397203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214384-93-6
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.147
MDL Number MFCD13185681
InChI Key UOBKAUNYVVOXMY-UHFFFAOYSA-N
Synonym 3-methoxy-2-trifluoromethyl benzoic acid
PubChem CID 73012640
IUPAC Name 3-methoxy-2-(trifluoromethyl)benzoic acid
SMILES COC1=CC=CC(=C1C(F)(F)F)C(=O)O

Specifications

Melting Point 161°C to 164°C
Color Cream
Quantity 1 g
Formula Weight 220.15
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 3-Methoxy-2-(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.