Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxycyclohexene 97.0+%, TCI America™
SDP

Catalog No. M24555ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24555ML 5 mL
M24551ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24555ML Supplier TCI America Supplier No. M24555ML

Chemical Identifiers

CAS 2699-13-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
InChI Key OKDKFTKUXADLSJ-UHFFFAOYSA-N
PubChem CID 137685
IUPAC Name 3-methoxycyclohexene
SMILES COC1CCCC=C1

Specifications

Color Colorless
Boiling Point 133°C
Quantity 5 mL
UN Number 3271
Formula Weight 112.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxycyclohexene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.