Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxypropionic acid, 97+%

Catalog No. p-7077416
Click to view available options
Quantity:
10 mL
50 mL

CAS: 2544-06-1 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2544-06-1
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
InChI Key YSIKHBWUBSFBRZ-UHFFFAOYSA-N
PubChem CID 134442
IUPAC Name 3-methoxypropanoic acid
SMILES COCCC(=O)O

Specifications

Density 1.1080g/mL
Boiling Point 116.0°C (9.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 10 mL
Specific Gravity 1.108
Synonym 3-methoxypropionic acid, propanoic acid, 3-methoxy, propanoic acid, methoxy, m-peg1-acid, propanoic acid,methoxy, acmc-1aiwk, 3-methoxy-propionic acid, #, propanoic acid,3-methoxy, .beta.-methoxypropionic acid
Formula Weight 104.11
Percent Purity 97+%
Chemical Name or Material 3-Methoxypropionic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.