Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxyquinoline-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. AAH5421103
Click to view available options
Quantity:
1 g

Catalog No. AAH5421103

Supplier: Thermo Scientific Chemicals H5421103

Only null left
Add to Cart

3-Methoxyquinoline-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxyquinoline-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 881656-35-5
Molecular Formula C16H20BNO3
Molecular Weight (g/mol) 285.15
MDL Number MFCD20527212
InChI Key MGEKTRDPBDREKY-UHFFFAOYSA-N
Synonym 3-methoxyquinoline-5-boronic acid pinacol ester, 3-methoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-quinoline, 3-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 3-methoxy-5-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl-quinoline
PubChem CID 68783972
IUPAC Name 3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
SMILES COC1=CN=C2C=CC=C(B3OC(C)(C)C(C)(C)O3)C2=C1

Specifications

Quantity 1 g
Formula Weight 285.15
Percent Purity 96%
Chemical Name or Material 3-Methoxyquinoline-5-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.