Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-butanol, ACS, 98.5+%

Catalog No. AA36716AP
Click to view available options
Quantity:
500 mL
1 L
4 L

Catalog No. AA36716AP

Supplier: Thermo Scientific Chemicals 036716AP

Only null left
Add to Cart

CAS: 123-51-3 | C5H12O | 88.15 g/mol

3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac's reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Melting Point °C to 117°C
Density 0.809
Boiling Point 130°C to 132°C
Flash Point 45°C (113°F)
Assay Percent Range 98.5+%
Refractive Index 1.406
Quantity 500 mL
UN Number UN1105
Beilstein 1718835
Merck Index 14,5195
Solubility Information Miscible with water,alcohol,ether,benzene,chloroform,petroleum ether,glacial acetic acid,acetone and oils.
Formula Weight 88.15
Percent Purity ≥98.5%
Grade ACS
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-butanol
Show More Show Less
Hazard Category H226-H315-H319-H332-H335
Hazard Statement GHS H Statement
H226-H332-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 204-633-5
RTECSNumber EL5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.