Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Methyl-1-butanol, ACS reagent
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -70.0°C |
| Density | 0.8000g/mL |
| Boiling Point | 130.0°C |
| Flash Point | 45°C |
| Packaging | Glass bottle |
| Refractive Index | 1.4050 to 1.4070 |
| Linear Formula | (CH3)2CHCH2CH2OH |
| Quantity | 500 mL |
| Residue after Evaporation | 0.003% max. |
| Beilstein | 01, 392 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Flammable liquid and vapour. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Repeated exposure may cause skin dryness or cracking. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. If skin |
| EINECSNumber | 204-633-5 |
| RTECSNumber | EL5425000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction