Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-butyne, 96%

Catalog No. p-7035713
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAH5343906

Supplier: Thermo Scientific Chemicals H5343906

Only null left
Add to Cart

CAS: 598-23-2 | C5H8 | 68.119 g/mol

3-Methyl-1-butyne is a terminal alkyne that under goes hydrosilylation reaction using phenylsilane. 3-Methyl-1-butyne is used as an intermediate for pharmaceuticals, in cosmetics and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-butyne is a terminal alkyne that under goes hydrosilylation reaction using phenylsilane. 3-Methyl-1-butyne is used as an intermediate for pharmaceuticals, in cosmetics and in chemical research.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Store in cool place. Light and heat sensitive. Incompatible with strong oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-23-2
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00039853
InChI Key USCSRAJGJYMJFZ-UHFFFAOYSA-N
Synonym 3-methyl-1-butyne, 1-butyne, 3-methyl, isopentyne, isopropylacetylene, isopropyl acetylene, 3-methylbutyne, 2-methyl-3-butyne, 3-methyl-but-1-yne, 3-methyl,1-butyne
PubChem CID 69019
ChEBI CHEBI:87379
IUPAC Name 3-methylbut-1-yne
SMILES CC(C)C#C

Specifications

Density 0.666
Boiling Point 30°C
Flash Point −30°C (−22°F)
Refractive Index 1.374
Linear Formula (CH3)2CHC≡CH
Quantity 5 g
UN Number UN3295
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 68.12
Percent Purity 97%
Chemical Name or Material 3-Methyl-1-butyne
Show More Show Less
Hazard Category H224-H315-H319-H335
Hazard Statement GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 209-925-6
RTECSNumber PF9060500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.