Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Methyl-2-pentene (cis- and trans- mixture) 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 mL
Specifications
Specifications
Color | Colorless |
Quantity | 1 mL |
UN Number | 2288 |
Formula Weight | 84.16 |
Percent Purity | ≥99.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 3-Methyl-2-pentene (cis- and trans- mixture) |
Safety and Handling
EINECSNumber | (2)-0031 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction