Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Methylacetophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
585-74-0
C9H10O
134.18
FSPSELPMWGWDRY-UHFFFAOYSA-N
3'-methylacetophenone,1-m-tolyl ethanone,3-methylacetophenone,m-methylacetophenone,ethanone, 1-3-methylphenyl,acetophenone, m-methyl,methyl m-tolyl ketone,1-3-methylphenyl ethan-1-one,3-acetyltoluene,acetophenone, 3'-methyl
11455
1-(3-methylphenyl)ethanone
CC1=CC=CC(=C1)C(=O)C
585-74-0
C9H10O
134.18
FSPSELPMWGWDRY-UHFFFAOYSA-N
3'-methylacetophenone,1-m-tolyl ethanone,3-methylacetophenone,m-methylacetophenone,ethanone, 1-3-methylphenyl,acetophenone, m-methyl,methyl m-tolyl ketone,1-3-methylphenyl ethan-1-one,3-acetyltoluene,acetophenone, 3'-methyl
11455
1-(3-methylphenyl)ethanone
CC1=CC=CC(=C1)C(=O)C
Density 1.0100g/mL
Quantity 5g
Packaging Glass bottle
Color Colorless to Yellow
Flash Point 85°C
Infrared Spectrum Authentic
Specific Gravity 1.01
Boiling Point 218.0°C to 220.0°C
Refractive Index 1.529
Melting Point -9.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3C6H4COCH3
Beilstein 07,307
Formula Weight 134.18
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3′-Methylacetophenone
Show More Show Less

Warning

EINECSNumber : 209-561-8