Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylisoquinoline, 95%

Catalog No. p-4089568
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 1125-80-0 | C10H9N | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125-80-0
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00024139
InChI Key FVVXWRGARUACNW-UHFFFAOYSA-N
Synonym isoquinoline, 3-methyl, 3-methyl-isoquinoline, unii-4quo6nyb5j, 4quo6nyb5j, isoquinolone-3-methyl, 3-methyl-isoquinolin, 3-methyl isoquinoline, 3-methylisoquinoline, 5-20-07-00410 beilstein handbook reference, ksc176i1j
PubChem CID 14306
IUPAC Name 3-methylisoquinoline
SMILES CC1=CC2=CC=CC=C2C=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 63.0°C to 65.0°C
Boiling Point 251.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material 3-Methylisoquinoline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-412-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.